missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mercury(II) Chloride, 99%, Reagent Grade, Honeywell™
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Packaging:
Poly bottle
Description
- ≥95% for general laboratory use.
Specifications
Specifications
Boiling Point | 302°C |
Melting Point | 276°C |
Color | White |
Physical Form | Solid |
Quantity | 1 kg |
Linear Formula | HgCl2 |
UN Number | UN1624 |
Formula Weight | 271.5g/mol |
Percent Purity | 99% |
Odor | Odorless |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'