missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEOX2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA549810
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total MEOX2 protein.
Mesodermal transcription factor that plays a key role in somitogenesis and is required for sclerotome development. Activates expression of CDKN1A and CDKN2A in endothelial cells, acting as a regulator of vascular cell proliferation. While it activates CDKN1A in a DNA-dependent manner, it activates CDKN2A in a DNA-independent manner. May have a regulatory role when quiescent vascular smooth muscle cells reenter the cell cycle.Specifications
MEOX2 | |
Polyclonal | |
Unconjugated | |
MEOX2 | |
AI528662; GAX; Growth arrest-specific homeobox; homeobox protein MOX-2; MEOX2; mesenchyme homeo box 2 (growth arrest-specific homeo box); mesenchyme homeobox 2; Mox2; Mox-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
17286, 29279, 4223 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P32443, P39020, P50222 | |
MEOX2 | |
Synthesized peptide derived from internal of human MEOX2. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |