missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ Meker-Fisher Burner

Bochem™ Meker-Fisher Burner regulates gas with a swivel screw.

Brand:  Bochem™ 7520

Code : A0

Additional Details : Weight : 0.80000kg

Product Code. 10142891

  • £81.05 / Each
Estimated Shipment: 22-07-2024
to see stock.
Add to basket

Description

Description

  • Air regulation
  • Gas regulation with swivel screw
Specifications

Specifications

Meker-Fisher Burner
Nickel, Steel
30 mm (Head), 80 mm (Base)
1300°C
Multigas
5 to 100 mbar, 3KW
200 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers