Learn More
MDH1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA555573
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: AGQIAYSLLY SIGNGSVFGK DQPIILVLLD ITPMMGVLDG VLMELQDCAL PLLKDVIATD KEDVAFKDLD VA Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 97%.
MDH1 is localized to the cytoplasm and may play pivotal roles in the malate-aspartate shuttle that operates in the metabolic coordination between cytosol and mitochondria.Specifications
MDH1 | |
Polyclonal | |
Unconjugated | |
MDH1 | |
B230377B03Rik; Cytosolic malate dehydrogenase; D17921; Diiodophenylpyruvate reductase; epididymis secretory protein Li 32; HEL-S-32; malate dehydrogenase (EC 1.1.1.37); malate dehydrogenase 1; malate dehydrogenase 1, NAD (soluble); malate dehydrogenase, cytoplasmic; malate dehydrogenase, cytoplasmic; malate dehydrogenase, peroxisomal; malate dehydrogenase, soluble; Malate dehydrogenase-like enzyme; Mdh; Mdh1; MDHA; Mdhl; MDH-s; MDL1; MGC:1375; Mor2; Mor-2; soluble malate dehydrogenase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4190 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P40925 | |
MDH1 | |
Recombinant protein corresponding to Human MDH1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only