missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Atago™ MASTER-SUR/Nα Model Clinical Refractometer
POA
Specifications
Type | Clinical Refractometer |
---|---|
ScaleRange | 1.000 to 1.060 Urine Specific Gravity, 0 to 12 g/100 mL Serum Protein |
IP Rating | IP65 |
Weight (Metric) | 110 g |
Specifications
Clinical Refractometer | |
1.000 to 1.060 Urine Specific Gravity, 0 to 12 g/100 mL Serum Protein | |
IP65 | |
110 g |