missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAPK4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | MAPK4 |
---|---|
Concentration | 0.22 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16838624
|
Proteintech
18738-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16828624
|
Proteintech
18738-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mitogen-activated protein kinase 4 is a member of the mitogen-activated protein kinase family. Tyrosine kinase growth factor receptors activate mitogen-activated protein kinases which then translocate into the nucleus where it phosphorylates nuclear targets.Specifications
MAPK4 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
P31152, Q63454, Q6P5G0 | |
225724, 54268, 5596 | |
Peptide | |
Primary | |
-20°C | |
MAPK4 |
0.22 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ERK 4, ERK3, Erk4, MAP kinase 4, MAP kinase isoform p63, MAPK 4, MAPK4, p63 MAPK, p63MAPK, PRKM4 | |
Mapk4 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |