missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MAP3K11 (MLK3), GST Tag Recombinant Protein
Human MAP3K11 (MLK3), GST Tag Recombinant Protein
Brand: Invitrogen™ PV3788
Code : 50
Additional Details : Weight : 0.05250kg
Description
MAP3K11 (MLK3) is a serine⁄threonine kinase that activates both the p38 MAP kinase and c-Jun kinase pathways.Specifications
NP_002410.1 | |
4296, 6258 | |
10 μg | |
MLK Kinase Family | |
Recombinant | |
Storage Buffer: 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Ser⁄Thr Kinases: TKL Group | |
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Omnia™ Bulk Substrate | |
mitogen-activated protein kinase kinase kinase 11 | |
Kinases & Inhibitors | |
Liquid | |
Immunology, Oncology, Signal Transduction |
Kinase Assay | |
120.1 kDa | |
MAP3K11 (MLK3) | |
Human | |
GST Tag | |
Biochemical terbium phospho-specific activity, Biochemical binding/kinetic | |
Dry Ice | |
MAP3K11 | |
Baculovirus | |
Recombinant, Full Length | |
Serine/Threonine Kinases |