missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAML3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA558073
This item is not returnable.
View return policy
Description
Immunogen sequence: GVGMVSGFGQ SMLVNSAITQ QHPQMKGPVG QALPRPQAPP RLQSLMGTVQ QGAQSWQQRS LQGMPGRTSG ELGPFNNGAS YPLQAGQPR Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 32%.
MAML3 acts as a transcriptional coactivator for NOTCH proteins. Has been shown to amplify NOTCH-induced transcription of HES1.Specifications
MAML3 | |
Polyclonal | |
Unconjugated | |
MAML3 | |
AV234550; BC049812; CAG repeat containing (glia-derived nexin I alpha); CAG repeat domain; CAGH3; ERDA3; expanded repeat domain, CAG/CTG 3; GDN; Glrp1; glutamine repeat protein 1; KIAA1816; MAM2; Mam-2; mam-3; MAML3; mastermind like 3 (Drosophila); mastermind like transcriptional coactivator 3; mastermind-2; mastermind-like 3; mastermind-like protein 3; mastermind-like transcriptional coactivator 3; mKIAA1816; polyglutamine rich; TNRC3; trinucleotide repeat containing 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55534 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96JK9 | |
MAML3 | |
Recombinant protein corresponding to Human MAML3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only