missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium acetate tetrahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals
£16.60 - £148.90
Chemical Identifiers
CAS | 16674-78-5 |
---|---|
Molecular Formula | C4H14MgO8 |
Molecular Weight (g/mol) | 214.45 |
MDL Number | MFCD00149214 |
InChI Key | XKPKPGCRSHFTKM-UHFFFAOYSA-L |
Synonym | magnesium acetate tetrahydrate, cromosa, acetic acid, magnesium salt, tetrahydrate, unii-i01g0ejc3b, i01g0ejc3b, magnesium diacetate tetrahydrate, magnesium diethanoate tetrahydrate, acmc-20aklu, magnesiumacetatetetrahydrate, ksc497c8p |
PubChem CID | 134717 |
IUPAC Name | magnesium(2+) diacetate tetrahydrate |
SMILES | O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10301071
|
Thermo Scientific Acros
423870050 |
5 g | Glass bottle |
£16.60
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10657031
|
Thermo Scientific Acros
423875000 |
500 g | Plastic bottle |
£53.80
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10637464
|
Thermo Scientific Acros
423870025 |
2.5 kg | Plastic bottle | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
16674-78-5 | |
214.45 | |
XKPKPGCRSHFTKM-UHFFFAOYSA-L | |
134717 | |
O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O |
C4H14MgO8 | |
MFCD00149214 | |
magnesium acetate tetrahydrate, cromosa, acetic acid, magnesium salt, tetrahydrate, unii-i01g0ejc3b, i01g0ejc3b, magnesium diacetate tetrahydrate, magnesium diethanoate tetrahydrate, acmc-20aklu, magnesiumacetatetetrahydrate, ksc497c8p | |
magnesium(2+) diacetate tetrahydrate |
Specifications
80.0°C | |
White | |
98.0 to 102% (Complexometry) | |
(CH3CO2)2Mg·4H2O | |
15,196 | |
magnesium acetate tetrahydrate, cromosa, acetic acid, magnesium salt, tetrahydrate, unii-i01g0ejc3b, i01g0ejc3b, magnesium diacetate tetrahydrate, magnesium diethanoate tetrahydrate, acmc-20aklu, magnesiumacetatetetrahydrate, ksc497c8p | |
XKPKPGCRSHFTKM-UHFFFAOYSA-L | |
magnesium(2+) diacetate tetrahydrate | |
134717 | |
≥98% | |
Authentic |
16674-78-5 | |
Crystalline Powder and/or Crystals | |
C4H14MgO8 | |
MFCD00149214 | |
15, 5718 | |
Solubility in water: 1200g/L (15°C). Other solubilities: soluble in alcohol | |
O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O | |
214.45 | |
214.46 | |
ACS Reagent | |
Magnesium acetate tetrahydrate |
Safety and Handling
HYGROSCOPIC
RUO – Research Use Only