missing translation for 'onlineSavingsMsg'
Learn More

Hellma Macro Cuvettes, 100-QS Quartz (pair)

Highly pure Suprasil™ quartz glass cuvettes with lids.

Brand:  Hellma 100-5-K-40

Code : NEW

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 15828881

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • High-purity synthetic fused silica quartz construction
  • UV/VIS range
  • Glass lids included
Specifications

Specifications

1750 μL
PTFE
Base Thickness: 1.5 mm
5 mm
45 mm
Suprasil Quartz, QS-100
9.5 mm
2/Pk
200-2500 nm
12.5 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers