missing translation for 'onlineSavingsMsg'
Learn More

m-Tolylmagnesium chloride, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Catalog Number p-4258691
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
121905-60-0, 109-99-9
C7H7ClMg
150.89
MFCD00238749
BVYCQPCLCZTSPD-UHFFFAOYSA-M
m-tolylmagnesium chloride, m-tolyl magnesium chloride, m-methylphenylmagnesium chloride, bvycqpclcztspd-uhfffaoysa-m, m-tolylmagnesium chloride solution 1.0 m in thf, m-tolylmagnesium chloride solution, 1.0 m in thf
4384483
CC1=CC([Mg]Cl)=CC=C1
This item is not returnable. View return policy
121905-60-0, 109-99-9
C7H7ClMg
150.89
MFCD00238749
BVYCQPCLCZTSPD-UHFFFAOYSA-M
m-tolylmagnesium chloride, m-tolyl magnesium chloride, m-methylphenylmagnesium chloride, bvycqpclcztspd-uhfffaoysa-m, m-tolylmagnesium chloride solution 1.0 m in thf, m-tolylmagnesium chloride solution, 1.0 m in thf
4384483
CC1=CC([Mg]Cl)=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 14.99, 82.64
CAS Max % 17.36, 85.01
Physical Form Solution
Quantity 100 mL
Flash Point −21°C
Solubility Information Solubility in water: reacts
IUPAC Name magnesium;methylbenzene;chloride
Formula Weight 150.89
Concentration or Composition (by Analyte or Components) 0.95 to 1.10M
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.956
Density 0.9560g/mL
Chemical Name or Material m-Tolylmagnesium chloride
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • m-Tolylmagnesium chloride, 1M solution in THF
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'