Learn More
Invitrogen™ Human LYN A, GST Tag Recombinant Protein
Human LYN A, GST Tag Recombinant Protein
Brand: Invitrogen™ PV6450
Code : NEW
Additional Details : Weight : 0.01000kg
Description
LYN is a protein-coding gene and is associated with diseases such as alexia and chorea-acanthocytosis. Activation of LYN has been shown to lead to imatinib resistance in CML patients. LYN B and the slightly shorter LYN A are the two isoforms of LYN. An important paralog of this gene is FRK. The full length LYN A protein enables researchers to study important mechanisms underlying disease.
- Increase disease relevance by interrogating a purified and active full-length LYN A reagent
- Uncover novel compounds or hits specific to the full length form
- Advance your research using the full-length protein and validated assays
- Assay Platforms: LanthaScreen™ TR-FRET Kinase Activity Assay, LanthaScreen™ Eu Kinase Binding Assay, and Z’-LYTE™ Kinase Activity Assay
Specifications
NP_002341.1 | |
81kDa | |
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws. If properly stored, this product is guaranteed for 6 months from date of purchase. Suspended in: 50 nM Tris-HCl, pH 7.5, 150 mM NaCl, 0.25 mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, 25% glycerol. |
|
JTK8, FLJ26625 | |
Human | |
GST Tag |
Kinase Assay, Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Biology, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation, Kinase Assay | |
1mg | |
Full Length | |
SRC Kinase Family | |
Recombinant | |
Insect |