Learn More
Invitrogen™ LXR-b Recombinant Human Protein, Ligand Binding Domain, GST-Tagged
LXR-b (Liver X Receptor beta) ligand binding domain is a 57.6 kDa recombinant human protein (amino acids 201-461) expressed as a GST fusion protein in baculovirus-infected insect cells.
Brand: Invitrogen™ PV4660
Code : 50
Additional Details : Weight : 0.50000kg
Description
LXR beta was expressed and purified in the absence of exogenous ligands, making it ideal for ligand binding and coregulator displacement studies.
Specifications
Nuclear Receptor Assay | |
50 μg | |
RUO - research use only | |
Liver X Receptor Beta | |
Recombinant | |
Hormone Receptors |
7376 | |
Store in ultra-cold freezer (-68 to -85°C). | |
Recombinant, Ligand Binding Domain | |
Human | |
GST Tag |