missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LTK (TYK1) Recombinant Human Protein

LTK (TYK1) is a member of the ros/insulin receptor family of kinases.

Brand:  Invitrogen™ PV4652

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15448743

  • £6900.00 / 1mg
Estimated Shipment: 06-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

LTK (TYK1) is involved in cell growth and differentiation.

Specifications

Specifications

NP_002335.2
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol
73.7kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Cytoplasmic
ALK Kinase Family
Recombinant
Baculovirus
Nuclear Receptor Assay
4058
1mg
RUO - research use only
LTK (TYK1)
Human
GST Tag
Frozen
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers