missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LTK (TYK1) Recombinant Human Protein
LTK (TYK1) is a member of the ros/insulin receptor family of kinases.
Brand: Invitrogen™ PR8108A
Code : NEW
Additional Details : Weight : 0.01000kg
Description
LTK (TYK1) is involved in cell growth and differentiation.
Specifications
NP_002335.2 | |
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol | |
100μg | |
RUO - research use only | |
LTK (TYK1) | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
73.7kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Cytoplasmic | |
ALK Kinase Family | |
Recombinant | |
Baculovirus |