missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Liquid Scintillation Glass Vials

Maintain the integrity of seals with a variety of cap liners. Wheaton™ Liquid Scintillation Glass Vials are manufactured from borosilicate glass.

Brand:  DWK Life Sciences 986582

Additional Details : Weight : 10.06970kg

 View more versions of this product

Product Code. 11380480

  • £316.00 / Pack of 500
Estimated Shipment: 21-06-2024
to see stock.
Add to basket

Description

Description

  • Foamed polyethylene liners resilient to acids, alkalis, solvents, alcohols, oils, household cosmetics and aqueous products
  • Metal foil/cork liners resilient to hydrocarbons, oils, ketones and alcohols
  • Polyethylene cone liners are crack resistant and offer superior torque retention and excellent sealing characteristics
Specifications

Specifications

Metal Foil
28 mm
20 mL
Urea
500 Pk.
Clear
Borosilicate Glass
500
61 mm
22-400
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers