missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Liquid Scintillation Glass Vials

Maintain the integrity of seals with a variety of cap liners. Wheaton™ Liquid Scintillation Glass Vials are manufactured from borosilicate glass.

Brand:  DWK Life Sciences 986580

Additional Details : Weight : 9.52350kg

 View more versions of this product

Product Code. 10523614

  • £358.00 / Pack of 500
Estimated Shipment: 08-10-2024
to see stock.
Add to basket

Description

Description

  • Foamed polyethylene liners resilient to acids, alkalis, solvents, alcohols, oils, household cosmetics and aqueous products
  • Metal foil/cork liners resilient to hydrocarbons, oils, ketones and alcohols
  • Polyethylene cone liners are crack resistant and offer superior torque retention and excellent sealing characteristics
Specifications

Specifications

Foamed Polyethylene
28 mm
20 mL
61 mm
22-400
Clear
Borosilicate Glass
Polypropylene
500 Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers