Learn More
Invitrogen™ LRRK2 [R1441C] Human Protein
LRRK2 is a multidomain protein that includes a kinase domain, a GTPase domain, and a leucine-rich repeat (LRR) domain.
Brand: Invitrogen™ PR8752A
Code : NEW
Additional Details : Weight : 0.00900kg
Description
Mutations in this gene have been associated with Parkinson 's disease, including the R1441C mutation in the GTPase domain.
Specifications
NP_940980.2 | |
204.8kDa | |
Kinase proteins are shipped on dry ice and should be stored at -80°C. | |
Catalytic | |
LRRK Kinase Family | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
100μg | |
RUO - research use only | |
LRRK2 R1441C | |
Human | |
Baculovirus |