missing translation for 'onlineSavingsMsg'
Learn More

Human LRRK2 [G2019S], GST Tag Recombinant Protein

LRRK2 is a multidomain protein that includes a kinase domain, a GTPase domain, and a leucine-rich repeat (LRR) domain.

Brand:  Invitrogen PR8764C

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15468683

  • £2535.00 / 100µg
Estimated Shipment: 03-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Mutations in the LRRK2 gene have been associated with Parkinson's disease. The G2019S mutation of LRRK2 is associated with an increase in catalytic activity.

Specifications

Specifications

NP_940980.2, G2019S
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Polysorbate 20, 2mM DTT, 50% glycerol
100μg
RUO - research use only
LRRK2 G2019S
Human
GST Tag
Frozen
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology
204.9kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Catalytic
LRRK Kinase Family
Recombinant
Baculovirus
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers