Learn More
Human LRRK2 [G2019S], GST Tag Recombinant Protein
LRRK2 is a multidomain protein that includes a kinase domain, a GTPase domain, and a leucine-rich repeat (LRR) domain.
Brand: Invitrogen PV4881
Code : 50
Additional Details : Weight : 0.50000kg
Description
Mutations in the LRRK2 gene have been associated with Parkinson's disease. The G2019S mutation of LRRK2 is associated with an increase in catalytic activity.
Specifications
NP_940980.2, G2019S | |
120892 | |
10μg | |
LRRK Kinase Family | |
Human | |
GST Tag | |
Biochemical ADP accumulation, Biochemical terbium phospho-specific activity, Biochemical binding/kinetic | |
Dry Ice | |
LRRK2 G2019S | |
Baculovirus | |
Recombinant, Catalytic, Mutant | |
Serine/Threonine Kinases |
Kinase Assay | |
204.9 kDa | |
LRRK2 G2019S | |
aa 970-2527; G2019S | |
Recombinant | |
Storage Buffer: 50 mM Tris pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Polysorbate 20, 2 mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Ser⁄Thr Kinases: TKL Group | |
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Adapta™ | |
Leucine-rich repeat kinase 2 | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction, Neurobiology |