missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRRK2 [D1994A] Human Protein
LRRK2 is a multidomain protein that includes a kinase domain, a GTPase domain, and a leucine-rich repeat (LRR) domain. Certain mutations (e.g. I2020T, G2019S, and R1441C) in this gene have been associated with Parkinson 's disease.
£2405.00 - £11360.00
Specifications
Accession Number | NP_940980.2 |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Molecular Weight (g/mol) | 205kDa |
Storage Requirements | Kinase proteins are shipped on dry ice and should be stored at -80°C. |
Regulatory Status | RUO - research use only |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15448713
|
Invitrogen™
PR9102A |
100μg |
£2405.00
100µg |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15498803
|
Invitrogen™
PV6052 |
1mg |
£11360.00
1mg |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
D1994A is an artificial mutation that renders the kinase catalytically inactive and thus serves as a useful control for activity assays, especially when testing novel substrates.
Specifications
NP_940980.2 | |
205kDa | |
RUO - research use only | |
LRRK2 D1994A | |
Human | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
Kinase proteins are shipped on dry ice and should be stored at -80°C. | |
Catalytic | |
LRRK Kinase Family | |
GST Tag | |
Frozen |