missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Loxistatin
£61.50 - £267.00
Description
A cell-permeable, irreversible inhibitor of cysteine proteases
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
88321-09-9 | |
C17H30N2O5 | |
EST; E-64d |
Specifications
White | |
A cell-permeable, irreversible inhibitor of cysteine proteases | |
C17H30N2O5 | |
EST; E-64d | |
Powder | |
CCOC(=O)[C@H]1O[C@@H]1C(=O)N[C@@H](CC(C)C)C(=O)NCCC(C)C | |
342.44 |
-30°C to -10°C | |
88321-09-9 | |
342.43 | |
Soluble in DMSO, DMF or ethanol | |
SRVFFFJZQVENJC-IHRRRGAJSA-N | |
ethyl (2S,3S)-3-{[(1S)-3-methyl-1-[(3-methylbutyl)carbamoyl]butyl]carbamoyl}oxirane-2-carboxylate |
Safety and Handling
Recommended Storage : Store at -20°C
RUO – Research Use Only