missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Powder Pack™ Reagents: Copper; 10mL

Pre-measured, unit dose reagent for water quaility testing

Brand:  Lovibond™ 530300

Code : NEW

Additional Details : Weight : 0.30000kg

Product Code. 15371655

  • £40.10 / Pack of 100
Estimated Shipment: 19-06-2024
to see stock.
Add to basket

Description

Description

  • Simply tear open Powder Pack and pour reagent into sample
  • Large design makes them easier to handle
  • “Easy Open” notch makes it simple to tear open the package
  • Easily create a “V”, to allow user to pour reagent into sample
  • Beveled edge prevents reagent from getting stuck in the corners

Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing

Specifications

Specifications

US EPA
10 mL
Test kits, colorimeters and spectrophotometers
Bicinchoninate
0.33 oz.
Dipotassium Hydrogen Orthophosphate 40 to 50%, Potassium Dihydrogenorthophosphate 40 to 50%, Sodium Ascorbate 10 to 20%, 2,2 Bichinolin-4,4'-dicarbon Acid Dipotassium trihydrate 95%
Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing
0.05 to 5mg/L
Copper Reagent
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers