missing translation for 'onlineSavingsMsg'
Learn More

Lithium trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals

Catalog Number p-2691664
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
33454-82-9
CF3LiO3S
156.01
MFCD00013227
MCVFFRWZNYZUIJ-UHFFFAOYSA-M
lithium trifluoromethanesulfonate, lithium triflate, li triflate, lithium trifluoromethanesulphonate, methanesulfonic acid, trifluoro-, lithium salt, unii-1c966kv50i, trifluoromethanesulfonic acid lithium salt, lithium 1+ ion trifluoromethanesulfonate, methanesulfonic acid, 1,1,1-trifluoro-, lithium salt 1:1, trifluoromethanesulfonic acid, lithium salt
3664839
lithium;trifluoromethanesulfonate
[Li+].C(F)(F)(F)S(=O)(=O)[O-]
This item is not returnable. View return policy
33454-82-9
CF3LiO3S
156.01
MFCD00013227
MCVFFRWZNYZUIJ-UHFFFAOYSA-M
lithium trifluoromethanesulfonate, lithium triflate, li triflate, lithium trifluoromethanesulphonate, methanesulfonic acid, trifluoro-, lithium salt, unii-1c966kv50i, trifluoromethanesulfonic acid lithium salt, lithium 1+ ion trifluoromethanesulfonate, methanesulfonic acid, 1,1,1-trifluoro-, lithium salt 1:1, trifluoromethanesulfonic acid, lithium salt
3664839
lithium;trifluoromethanesulfonate
[Li+].C(F)(F)(F)S(=O)(=O)[O-]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 423.0°C
Color White or Colorless
Infrared Spectrum Authentic
Assay Percent Range 4.34 to 4.56% (Li)
Packaging Glass bottle
Linear Formula F3CSO3Li
Quantity 5 g
Solubility Information Solubility in water: soluble
Formula Weight 156.01
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Lithium trifluoromethanesulfonate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Lithium trifluoromethanesulfonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'