missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium dihydrogen phosphate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
Description
Employed as a precursor for LiMX2O7 (M = Fe, V; x =P, As) cathode materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Employed as a precursor for LiMX2O7 (M = Fe, V; x =P, As) cathode materials.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly sealed and store
Employed as a precursor for LiMX2O7 (M = Fe, V; x =P, As) cathode materials.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly sealed and store
Specifications
Melting Point | >100°C |
CAS | 13453-80-0 |
Quantity | 250 g |
Molecular Formula | H2LiO4P |
MDL Number | MFCD00016188 |
Synonym | lithium dihydrogen phosphate, monolithium phosphate, lithium dihydrophosphate, lithium dihydrogenorthophosphate, lithium dihydrogen orthophosphate, phosphoric acid, monolithium salt, lithium phosphate monobasic, phosphoric acid, lithium salt 1:1, lithiumdihydrogenphosphate, li.h2o4p |
Solubility Information | Slightly soluble in water. |
InChI Key | SNKMVYBWZDHJHE-UHFFFAOYSA-M |
SMILES | [Li+].OP(=O)(O)[O-] |
IUPAC Name | lithium;dihydrogen phosphate |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'