missing translation for 'onlineSavingsMsg'
Learn More

Lithium Acetate Dihydrate, purum p.a, ≥97.0% (NT), Honeywell™

Product Code. 15656040
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Purum p.a., crystallized,≥97.0% (NT)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6108-17-4
Molecular Formula C2H7LiO4
Molecular Weight (g/mol) 102.014
MDL Number MFCD00066949
InChI Key IAQLJCYTGRMXMA-UHFFFAOYSA-M
Synonym lithium acetate dihydrate, lithiumacetatedihydrate, lithium acetate dihydrate, reagent grade, lioac-2h2o, lioac.2h2o, ksc498a1d, c2h3o2.li.2h2o, lithotab acetate ion dihydrate, acetic acid lithium salt dihydrate
PubChem CID 23666338
IUPAC Name lithium;acetate;dihydrate
SMILES [Li+].CC(=O)[O-].O.O

Specifications

Melting Point 53°C to 56°C
Color White
Physical Form Solid
Quantity 1 kg
Linear Formula CH3COOLi · 2H2O
UN Number NONH for all modes of transport
Beilstein 3564320
Formula Weight 102.02g/mol
Percent Purity ≥97.0% (NT)
Odor No data
Packaging Poly bottle
Density 1.3g/cm3
Chemical Name or Material Lithium acetate dihydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'