Learn More
Liqui-Gel 37.5:1, MP Biomedicals
A 40% (w/v) solution of Ultra Pure Acrylamide (38.96%) and bis-Acrylamide (1.04%) in specially-prepared deionized water. Final ratio is 37.5:1
Brand: MP Biomedicals 04800804
Code : NEW
Additional Details : CAS Number : 79-06-1 Weight : 0.50000kg Transport : UN number : 3426 Chem class : 6.1 Pack group : III
Specifications
Liqui-Gel™ 37.5:1 | |
500 mL |
Safety and Handling
- liqui-gel 37:5:1
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 2
- Germ cell mutagenicity Category 1B
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 1
Hazard Statement
- H301-Toxic if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H361f-Suspected of damaging fertility.
- H372-Causes damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P262-Do not get in eyes, on skin, or on clothing.
- P263-Avoid contact during pregnancy/while nursing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : acrylamide, N'N-methylenediacrylamide