missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LIMK2 Recombinant Human Protein
LIMK2 is a serine/threonine kinase.
Brand: Invitrogen™ PR7519B
Code : NEW
Additional Details : Weight : 0.01000kg
Description
LIMK2 is involved in regulation of actin cytoskeletal changes via phosphorylation of cofilin. LIMK2 has also been implicated in brain development.
Specifications
NP_005560 | |
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol | |
100μg | |
RUO - research use only | |
LIMK2 | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
99.7kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
LISK Kinase Family | |
Recombinant | |
Baculovirus |