missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human LIMK1, His Tag Recombinant Protein
£1604.00 - £6900.00
Specifications
Accession Number | NP_002305 |
---|---|
For Use With (Application) | Kinase Assay |
Molecular Weight (g/mol) | 44.7 kDa |
Gene Alias | LIMK1, LIMK |
KinaseFamily | LISK Kinase Family |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15498623
|
Invitrogen™
PR7844A |
100μg |
£1604.00
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10524473
|
Invitrogen™
PV4348 |
1 mg |
£6900.00
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
LIMK1 is involved in regulation of actin cytoskeletal changes via phosphorylation of cofilin. LIMK1 has also been implicated in brain development.
Specifications
NP_002305 | |
44.7 kDa | |
LISK Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Baculovirus |
Kinase Assay | |
LIMK1, LIMK | |
Human | |
His Tag | |
Dry Ice |