Learn More
BINDER™ Light Photometer
For Use With: BINDER units KBW 240, KBW 400, KBW 720
Brand:  BINDER™ 8012-0910
Product Code. 15671966
  • £1850.00 / Each
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • 25 measuring points on 3 measurement levels
  • Spectral distribution of either 385 to 785 nm or 250 to 785 nm
  • For illumination, irradiance, visible light, and UV-A (8012-9010 only)
Specifications
Specifications
Light photometry - including certificate, illumination and irradiance for visible light and UV-A, 25 measuring points on 3 measurement levels, as well as spectral distribution (250 - 785 nm)
25 Measurement Points
Illumination, Irradiance, Visible Light, UV-A
BINDER Chambers KBW 240, KBW 400, KBW 720
Light Photometer
Certificate