missing translation for 'onlineSavingsMsg'
Learn More

Crowthorne Hi-Tech Services Light box 230V 50Hz for F100

Brand:  Crowthorne Hi-Tech Services LA-F100RL

Light box 230V 50Hz for F100

Code : 04

Additional Details : Weight : 20.00000kg

Product Code. 12841710

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers