missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ LDPE Wash Bottles with Venting Valve
High-quality wash bottles with integrated wash stream and sealing cone closure.
£36.09 - £52.63
Specifications
Material | LDPE |
---|---|
Closure Included | Yes |
Mouth | Wide |
Product Type | Wash Bottle |
Closure Type | Stopper |
Product Code | Brand | Capacity (Metric) | Chemical Label | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Chemical Label | Price | Quantity & Availability | ||||||
10325281
|
BRAND™
144022 |
250 mL | Ethanol | N/A | N/A | N/A | |||||
10401931
|
BRAND™
144020 |
250 mL | Methanol | N/A | N/A | N/A | |||||
10709201
|
BRAND™
144024 |
500 mL | Distilled Water | N/A | N/A | N/A | |||||
10439081
|
BRAND™
144026 |
500 mL | Methanol | N/A | N/A | N/A | |||||
10761383
|
BRAND™
144028 |
500 mL | Ethanol | N/A | N/A | N/A | |||||
10247781
|
BRAND™
144027 |
500 mL | Isopropanol | N/A | N/A | N/A | |||||
10489261
|
BRAND™
144025 |
500 mL | Acetone | N/A | N/A | N/A | |||||
Description
- For distilled water and organic solvents only
- Large opening for convenient filling
- Label with designation of contents and general safety statements in English (was CAS numbers and NFPA codes) and German (was CAS numbers)
- Manufactured from flexible PE-LD, limited to temperatures below 80°C
Specifications
LDPE | |
Wide | |
Stopper |
Yes | |
Wash Bottle |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BRAND™ LDPE Wash Bottles with Venting Valve