missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Lck, His Tag Recombinant Protein
LCK Recombinant Human Protein
£437.00 - £6800.00
Specifications
Accession Number | NP_005347 |
---|---|
Molecular Weight (g/mol) | 60.6 kDa |
Gene Alias | LCK |
Species | Human |
Protein Tag | His Tag |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10544443
|
Invitrogen™
P3043 |
10 μg |
£437.00
Each |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10073142
|
Invitrogen™
PV4081 |
1mg |
£6800.00
1mg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15468393
|
Invitrogen™
PR4170A |
100 μg |
£1574.00
100µg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
LCK is a SRC-related kinase family expressed in lymphocytes.Specifications
NP_005347 | |
LCK | |
His Tag |
60.6 kDa | |
Human | |
Baculovirus |
For Research Use Only. Not for use in diagnostic procedures.