missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lanthanum(III) chloride hydrate, Honeywell™
∼7 mol/mol water,≥98.5% (calc. based on dry substance, AT)
£89.37 - £468.51
Chemical Identifiers
CAS | 20211-76-1 |
---|---|
Molecular Formula | Cl3La |
Molecular Weight (g/mol) | 245.26 |
MDL Number | MFCD00149753 |
InChI Key | ICAKDTKJOYSXGC-UHFFFAOYSA-K |
IUPAC Name | lanthanum(3+) trichloride |
SMILES | [Cl-].[Cl-].[Cl-].[La+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15616040
|
Honeywell Research Chemicals
61490-100G |
100 g | N/A | ||||||
15626040
|
Honeywell Research Chemicals
61490-500G |
500 g |
£468.51
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Chemical Identifiers
20211-76-1 | |
245.26 | |
ICAKDTKJOYSXGC-UHFFFAOYSA-K | |
[Cl-].[Cl-].[Cl-].[La+3] |
Cl3La | |
MFCD00149753 | |
lanthanum(3+) trichloride |
Specifications
20211-76-1 | |
LaCl3 · xH2O | |
NONH for all modes of transport | |
[Cl-].[Cl-].[Cl-].[La+3] | |
245.26 | |
≥98.5% (calculated based on dry substance, AT) |
Cl3La | |
MFCD00149753 | |
ICAKDTKJOYSXGC-UHFFFAOYSA-K | |
lanthanum(3+) trichloride | |
245.26g/mol (anhydrous basis) | |
Lanthanum(III) chloride hydrate |
Safety and Handling
P261-P305 + P351 + P338
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title