Learn More
Lancer™ Laboratory Machine Detergent
Use in both Lancer and other quality washing machines. Lancer™ Laboratory Machine Detergent has been specially formulated to give the best possible results.
Brand: Lancer™ LCD25
This item is not returnable.
View return policy
Description
- Chemical cleaning compound
- Health and Safety Data Sheets available to comply with CHP regulations
Specifications
Laboratory Machine Detergent | |
Formulated to give the best possible results in both Lancer and other quality washing | |
25 L |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Carcinogenicity Category 2
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H351-Suspected of causing cancer.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P264-Wash thoroughly after handling.
- P405-Store locked up.
Supplemental information
- MIXTURE LIST-Contains : Trisodium nitrilotriacetate, Sodium hydroxide