Learn More
Labconco™ HEPA Filter for Rotary Vane Vacuum Pump and FreeZone™ Freeze Dry Systems
Fits between freeze dryer and pump to prevent entry of particulate contaminants into pump
Brand:  Labconco™ 7386500
Additional Details : Weight : 0.50000kg
Product Code. 15730178
  • £695.00 / Each
Estimated Shipment: 29-08-2024
to see stock.
Description
Description
HEPA Filter for all Labconco FreeZone Freeze Dry System when connecting to Labconco Rotary Vane or Hybrid Vacuum Pumps. Filter fits between the freeze dryer and pump to prevent particulate contaminants from getting into the pump.
Specifications
Specifications
1 lb.