missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ General Glove Box: Oxygen and Moisture Removal Column

Accessories for Labconco General Glove Box

Brand:  Labconco™ 5244200

Additional Details : Weight : 0.80000kg

Product Code. 11781459

  • £2775.00 / Each
Estimated Shipment: 04-11-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • For use with Labconco General Glove Box
  • Provides less than 5ppm oxygen and less than 50ppm moisture (refillable)

Specifications

Specifications

Precise™ Basic™ Glove Box and Controlled Atmosphere Glove Box
26 lb.
11.8 Kg
Oxygen, Moisture Removal Column
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers