Learn More
Labconco™ Blower Foot Switch for Precise™ Glove Boxes
Accessory for Precise Glove Boxes
Brand: Labconco™ 5241600
Code : NEW
Additional Details : Weight : 0.80000kg
Description
- Provides hands-free means to shut off power to the built-in blower
Specifications
Precise™ HEPA Filtered Glove Box, XPert™ Weigh Box | |
4.5 lb. | |
2 Kg | |
Blower Foot Switch |