missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ Ready-To-Use Streptavidin Peroxidase
Epredia™ Lab Vision Steptravidin Peroxidase (Ready-to-Use) reagent is intended for use in a labeled streptavidin-biotin immunoenzymatic antigen detection system.
£317.00
Specifications
Description | Streptavidin Peroxidase (Ready-To-Use) |
---|---|
Content And Storage | 2deg;C to 8°C. Each component is stable for 18 months. |
Product Line | Lab Vision™ |
Product Type | Streptavidin Peroxidase |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12694067
|
Epredia™
TS-060-HR |
60 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Storage Conditions: Store at 2°-8°C.Specifications
Streptavidin Peroxidase (Ready-To-Use) | |
2deg;C to 8°C. Each component is stable for 18 months. | |
Lab Vision™ | |
Streptavidin Peroxidase |