missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ p53 Rabbit Monoclonal Antibody (Clone Y5)

Employ part of a comprehensive antibody portfolio with the Epredia™ p53, Rabbit Monoclonal Antibody (Clone Y5).

Brand:  Epredia™ RM-2103-S

Code : LD

Additional Details : Weight : 0.00250kg

 View more versions of this product

Product Code. 12673607

  • £800.00 / 1mL
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

p53 is a tumor suppressor gene expressed in a wide variety of tissue types and is involved in regulating cell growth, replication, and apoptosis. It binds to mdm2, SV40 T antigen and human papilloma virus E6 protein. p53 senses DNA damage and possibly facilitaes repair. Mutation involving p53 is found in a wide variety of malignant tumors, including breast, ovarian, bladder, colon, lung, and melanoma. p53 is detected by immunostains only if the mutation causes protein stability (although protein may be nonfunctional) or a physiologic response. The physiologic response may be due to genetic changes induced by cellular stress from anoxia or aneuploidy; or genetic damage to other pathways controlling p53 such as MDM2 or alternative reading frame.

Host Species: Rabbit

Clone: Y5

Species Reactivity: Human.Others not known.

Immunogen: A synthetic peptide derived from N- terminal residues of human p53

Molecular Weight: 53kDa

Positive Control: Colon or Breast CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

p53
Monoclonal
Unconjugated
A synthetic peptide derived from N- terminal residues of human p53
IVD
Primary
Concentrated
Immunohistochemistry (Paraffin)
Y5
Rabbit
1 mL
Cancer and Tumor Biology
Human
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers