Learn More
L-Tyrosine disodium salt, 98%, Thermo Scientific Chemicals
£53.16 - £438.18
Chemical Identifiers
CAS | 69847-45-6 |
---|---|
Molecular Formula | C9H9NNa2O3 |
Molecular Weight (g/mol) | 225.16 |
MDL Number | MFCD00037203 |
InChI Key | ASIYFCYUCMQNGK-RIYJSUBBNA-L |
Synonym | sodium s-2-amino-3-4-hydroxyphenyl propanoate |
PubChem CID | 66602062 |
SMILES | [Na+].[Na+].N[C@@H](CC1=CC=C([O-])C=C1)C([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10326874
|
Thermo Scientific Acros
426410250 |
25 g | Glass Bottle |
£53.16
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10306734
|
Thermo Scientific Acros
426411000 |
100 g | Glass Bottle |
£125.63
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10755125
|
Thermo Scientific Acros
426415000 |
500 g | Glass Bottle |
£438.18
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
69847-45-6 | |
225.16 | |
ASIYFCYUCMQNGK-RIYJSUBBNA-L | |
66602062 |
C9H9NNa2O3 | |
MFCD00037203 | |
sodium s-2-amino-3-4-hydroxyphenyl propanoate | |
[Na+].[Na+].N[C@@H](CC1=CC=C([O-])C=C1)C([O-])=O |
Specifications
>300.0°C | |
98% | |
NaOC6H4CH2CH(NH2)CO2Na | |
sodium s-2-amino-3-4-hydroxyphenyl propanoate | |
[Na+].[Na+].N[C@@H](CC1=CC=C([O-])C=C1)C([O-])=O | |
− 12.90 | |
66602062 | |
98% |
69847-45-6 | |
C9H9NNa2O3 | |
MFCD00037203 | |
ASIYFCYUCMQNGK-RIYJSUBBNA-L | |
disodium (2S)-2-amino-3-(4-oxidophenyl)propanoate | |
225.16 | |
225.15 | |
L-Tyrosine disodium salt |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
- L-Tyrosine disodium salt
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only