Learn More
Copan Diagnostics L-Shaped Spreaders
L-shaped spreaders
POA
Specifications
Material | Plastic |
---|---|
Sterility | Sterile |
For Use With (Equipment) | Culture Plates |
Shape | L-Shape |
For Use With (Application) | Designed for spreading liquid samples, enables even spreading of liquid samples across the surface of agar plates without gouging or cutting the medium |
Description
L-shaped spreaders
L-Shaped Plastic Spreaders
- For spreading liquid samples
- Foot of the spreader has a completely smooth, rounded surface, free of rough edges and imperfections in the plastic
- Enables even spreading of liquid samples across the surface of agar plates without gouging or cutting the medium
- Available in two sterile peel pack sizes, five or ten spreaders per pack
Specifications
Plastic | |
Culture Plates | |
Designed for spreading liquid samples, enables even spreading of liquid samples across the surface of agar plates without gouging or cutting the medium |
Sterile | |
L-Shape |