missing translation for 'onlineSavingsMsg'
Learn More

L-Dihydroorotic acid, 99%, Thermo Scientific Chemicals

£115.00 - £487.00

Chemical Identifiers

CAS 5988-19-2
Molecular Formula C5H6N2O4
Molecular Weight (g/mol) 158.11
InChI Key UFIVEPVSAGBUSI-REOHCLBHSA-N
Synonym l-dihydroorotic acid, s-2,6-dioxohexahydropyrimidine-4-carboxylic acid, l-dihydroorotate, dihydro-l-orotic acid, s-dihydroorotic acid, l-hydroorotic acid, s-dihydroorotate, hydroorotic acid, l, 4s-2,6-dioxo-1,3-diazinane-4-carboxylic acid, s-4,5-dihydroorotate
PubChem CID 439216
ChEBI CHEBI:17025
IUPAC Name (4S)-2,6-dioxo-1,3-diazinane-4-carboxylic acid
SMILES C1C(NC(=O)NC1=O)C(=O)O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
11906541
View Documents Promotion Details
Thermo Scientific Acros
437240010
1 g Glass bottle
£115.00
1g
Please to purchase this item. Need a web account? Register with us today!
11916541
View Documents Promotion Details
Thermo Scientific Acros
437240050
5 g Glass bottle
£487.00
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5988-19-2
158.11
l-dihydroorotic acid, s-2,6-dioxohexahydropyrimidine-4-carboxylic acid, l-dihydroorotate, dihydro-l-orotic acid, s-dihydroorotic acid, l-hydroorotic acid, s-dihydroorotate, hydroorotic acid, l, 4s-2,6-dioxo-1,3-diazinane-4-carboxylic acid, s-4,5-dihydroorotate
CHEBI:17025
C1C(NC(=O)NC1=O)C(=O)O
C5H6N2O4
UFIVEPVSAGBUSI-REOHCLBHSA-N
439216
(4S)-2,6-dioxo-1,3-diazinane-4-carboxylic acid

Specifications

5988-19-2
White
98.5% min. (HPLC)
l-dihydroorotic acid, s-2,6-dioxohexahydropyrimidine-4-carboxylic acid, l-dihydroorotate, dihydro-l-orotic acid, s-dihydroorotic acid, l-hydroorotic acid, s-dihydroorotate, hydroorotic acid, l, 4s-2,6-dioxo-1,3-diazinane-4-carboxylic acid, s-4,5-dihydroorotate
C1C(NC(=O)NC1=O)C(=O)O
158.11
CHEBI:17025
99%
L-Dihydroorotic acid
254.0°C to 255.0°C
Authentic
C5H6N2O4
UFIVEPVSAGBUSI-REOHCLBHSA-N
(4S)-2,6-dioxo-1,3-diazinane-4-carboxylic acid
439216
158.11
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:

GHS Signal Word: Warning

Product Identifier
  • L-Dihydroorotic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only