Learn More
Ecolab™ Klercide™ Sporicidal Low Residue Peroxide
For use in control of most risks of contamination including difficult bacterial spores. Ecolab™ Klercide™ Sporicidal Low Residue Peroxide can be used in most cleanroom applications.
Brand: Ecolab™ 3079080
Code : NEW
Additional Details : CAS Number : 7722-84-1 Weight : 6.12000kg
Description
- Sterile cleanroom biocide
- Broad spectrum activity
- Used as part of rotation with any other Ecolab Contamination Control biocide
- Action point use to remove troublesome organisms
- Fully adjustable trigger spray allows liquid to be dispensed as either jet or spray
- Does not require any special disposal
- 0.2 micron filtered, aseptically filled and double bagged under Grade A laminar flow in a Grade B (ISO Class 5) cleanroom using strictly controlled and validated aseptic process
Certifications
Manufactured by a ISO 9001:2008, ISO 13485:2012 and European Medical Device Directive 93/42/EEC accredited company.
Specifications
Cleanroom Biocide | |
Blend of 6% hydrogen peroxide and deionised water | |
Disinfectant with sporicidal activity | |
1 L |
Sterile | |
Filtered | |
33.8 oz. |
Safety and Handling
- Klercide Sporicidal Low Residue Peroxide
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
- MIXTURE LIST-Contains : Hydrogen peroxide