Learn More
DWK Life Sciences Kimble&chase; Kimble™ Kontes™ ULTRA-WARE™ Filtration Adapter
This adapter assembly is designed to allow connection of a 47mm or 90mm filtration support base with a straight drip tip to a bottle or reservoir with a GL 45 thread.
Brand: DWK Life Sciences 179950-4532
Code : NEW
Additional Details : Weight : 0.50000kg
Description
- Filtrate is collected directly into a media-type bottle when using a standard membrane filtration assembly
- All components are steam-autoclavable
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Filtration Adapter Assembly | |
32 mm |