Learn More
DWK Life Sciences Kimble™ KIMAX™ Iodine Determination Flasks with PTFE Stopper
For analysis of dry shellac, varnishes, oils, fatty acids
POA
Specifications
Closure Material | PTFE |
---|---|
Material | Borosilicate Glass |
Writing Surface | Matte Finish Ceramic Enamel Marking |
Closure Size | 22 |
Description
For analysis of dry shellac, varnishes, oils, fatty acids
250mL size
- Analyze dry shellac and shellac varnishes according to ASTM™ D 29
- Determine total iodine value of oils, fatty acids, and bodied oils according to ASTM D 1541
- Test drying oils for use in paints and varnishes according to ASTM D 555
Compliance
Designed from ASTM E 1404, Type II, Class 3 requirements
Specifications
PTFE | |
Borosilicate Glass | |
Matte Finish Ceramic Enamel Marking | |
22 |