missing translation for 'onlineSavingsMsg'
Learn More

Ketoconazole, Fisher BioReagents

Product Code. 10032624
Click to view available options
Quantity:
50 mg
100 mg
500 mg
1 g
Packaging:
Amber Glass
This item is not returnable. View return policy
This item is not returnable. View return policy
Ketoconazole is a cytochrome P450 inhibitor and demonstrates anti-psoriatic activity. An antifungal agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65277-42-1
Molecular Formula C26H28Cl2N4O4
Molecular Weight (g/mol) 534.452
InChI Key XMAYWYJOQHXEEK-SIULDFEJSA-N
Synonym ketoconazole
PubChem CID 76973198
IUPAC Name 2,2,2-trideuterio-1-[4-[4-[[(2R,4S)-2-(2,4-dichlorophenyl)-2-(imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazin-1-yl]ethanone
SMILES CC(=O)N1CCN(CC1)C2=CC=C(C=C2)OCC3COC(O3)(CN4C=CN=C4)C5=C(C=C(C=C5)Cl)Cl

Specifications

Melting Point 148°C
Color White
Ignition Residue < 0.1%
Loss on Drying < 0.5%
Melting Range Inclusive between 148°C
Assay Percent Range 98%
Packaging Amber Glass
Quantity 500 mg
Merck Index 15, 5349
Formula Weight 531.44
Physical Form Solid
Chemical Name or Material Ketoconazole
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'