missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KAP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | KAP1 |
---|---|
Concentration | 0.4 mg/mL |
Applications | Immunocytochemistry, Flow Cytometry, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16854284
|
Proteintech
15202-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16844284
|
Proteintech
15202-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TIF1 beta is a member of the tripartite motif (TRIM) family. It is composed of a N-terminal RBCC (RING finger, B boxes, coiled coil) motif, and a C-terminal bromodomain which is preceded by a PHD finger. TIF1 beta has been reported to function as a co-repressor for Kruppel-associated box (KRAB)-domain-containing zinc finger proteins. As such, it plays a crucial role in early embryogenesis. It appears that the co-repressor function of TIF1 beta is related to its organization and/or maintece of higher-order, heterochromatin-like structures.Specifications
KAP1 | |
Immunocytochemistry, Flow Cytometry, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
O08629, Q13263, Q62318 | |
10155, 116698, 21849 | |
KAP1 Fusion Protein Ag7350 | |
Primary | |
-20°C | |
TRIM28 |
0.4 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
KAP 1, KAP1, KRAB associated protein 1, KRAB interacting protein 1, KRIP 1, Nuclear corepressor KAP 1, RING finger protein 96, RNF96, TF1B, TIF1 beta, TIF1B, TRIM28, tripartite motif containing 28 | |
TRIM28 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |