Learn More
3M™ Jupiter™ Powered Air Turbo Filters
Use with the Jupiter PAPR. The 3M Jupiter™ Powered Air Turbo Filters are available in a wide selection of gas or particulate protection.
Brand: 3M™ 7000145610
Code : NEW
Additional Details : Weight : 0.77400kg
Description
Filters are to be used with the Jupiter Powered Air Turbo respirator.Specifications
ABE1P3 | |
Combined Gas & Vapor and Particulate | |
Jupiter™ Powered-Air Respirator | |
1 Ea. |