Learn More
3M™ Jupiter™ Powered Air Turbo Filters
Use with the Jupiter PAPR. The 3M Jupiter™ Powered Air Turbo Filters are available in a wide selection of gas or particulate protection.
Brand: 3M™ 7000006599
87.40 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.92700kg
Description
Filters are to be used with the Jupiter Powered Air Turbo respirator.Specifications
A2BEK1P | |
Combined Gas & Vapor and Particulate | |
Jupiter™ Powered-Air Respirator | |
1 Ea. |